accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18593-44-7

IUPAC Name: 5,6-dimethyl-3H,4H-thieno[2,3-d]pyrimidin-4-one
Synonyms: 5,6-dimethyl-3H-thieno[2,3-d]pyrimidin-4-one
Molecular Weight (g/mol): 180.23
Molecular Formula: C8H8N2OS
InChi Key: QAFMGDDXMIKGEY-UHFFFAOYSA-N
SMILES: CC1=C(C)C2=C(S1)N=CNC2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results