accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 186000-41-9

IUPAC Name: 2-fluorobenzen-1-ide; iodozincylium
Synonyms: 2-fluorobenzen-1-ide; iodozincylium
Molecular Weight (g/mol): 287.38
Molecular Formula: C6H4FIZn
InChi Key: UEHRMZUVCBJTTI-UHFFFAOYSA-M
SMILES: [Zn+]I.FC1=CC=CC=[C-]1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results