Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triisopropylphosphonium Tetrafluoroborate

Catalog No. 698466500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-846-6500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 69-846-6500MG Supplier Sigma Aldrich Supplier No. 698466500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Synthesis of triisopropylphosphine complexes of ruthenium(II)Imine hydrogenationDihydride and alkene-++6-arene complexes

Specifications

CAS 186000-41-9
Density 1.03 g/mL at 25°C
Boiling Point 65°C
Molecular Formula C6H4FIZn
Linear Formula FC6H4ZnI
Quantity 500 mg
MDL Number MFCD02260162
Molecular Weight (g/mol) 287.39 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.