accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CH3CCH3OOOOOOH

CAS RN 186020-66-6

IUPAC Name: tert-butyl 3-{2-[2-(2-hydroxyethoxy)ethoxy]ethoxy}propanoate
Synonyms: tert-butyl 3-{2-[2-(2-hydroxyethoxy)ethoxy]ethoxy}propanoate
Molecular Weight (g/mol): 278.35
Molecular Formula: C13H26O6
InChi Key: KSXVEOLRERRELV-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)CCOCCOCCOCCO
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (3)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)