accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1861-40-1

IUPAC Name: N-butyl-N-ethyl-2,6-dinitro-4-(trifluoromethyl)aniline
Synonyms: benefin benfluralin
Molecular Weight (g/mol): 335.28
Molecular Formula: C13H16F3N3O4
InChi Key: SMDHCQAYESWHAE-UHFFFAOYSA-N
SMILES: CCCCN(CC)C1=C(C=C(C=C1[N+]([O-])=O)C(F)(F)F)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results