accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18618-64-9

IUPAC Name: pentadecan-8-amine
Synonyms: pentadecan-8-amine
Molecular Weight (g/mol): 227.44
Molecular Formula: C15H33N
InChi Key: NTPSQPZXFVRULZ-UHFFFAOYSA-N
SMILES: CCCCCCCC(N)CCCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results