Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

8-Pentadecanamine 96.0+%, TCI America™
SDP

Catalog No. P25435ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P25435ML 5 mL
P25431ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P25435ML Supplier TCI America Supplier No. P25435ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18618-64-9
Molecular Formula C15H33N
Molecular Weight (g/mol) 227.436
MDL Number MFCD00087197
InChI Key NTPSQPZXFVRULZ-UHFFFAOYSA-N
Synonym 1-Heptyloctylamine, 8-Aminopentadecane
PubChem CID 232907
IUPAC Name pentadecan-8-amine
SMILES CCCCCCCC(CCCCCCC)N

Specifications

Boiling Point 117°C
Quantity 5 mL
UN Number 2735
Formula Weight 227.44
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 8-Pentadecanamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.