accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSMgBr

CAS RN 18620-04-7

IUPAC Name: bromo[4-(methylsulfanyl)phenyl]magnesium
Synonyms: bromo[4-(methylsulfanyl)phenyl]magnesium
Molecular Weight (g/mol): 227.4
Molecular Formula: C7H7BrMgS
InChi Key: SSVQCUSLJVVRCA-UHFFFAOYSA-M
SMILES: CSC1=CC=C([Mg]Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results