Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Thioanisolemagnesium bromide, 0.5M solution in THF, AcroSeal™

Catalog No. AC399071000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC399071000 100 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC399071000 Supplier Thermo Scientific Chemicals Supplier No. 399071000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 18620-04-7 | C7H7BrMgS | 227.40 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18620-04-7 , 109-99-9
Molecular Formula C7H7BrMgS
Molecular Weight (g/mol) 227.40
MDL Number MFCD00672010
InChI Key SSVQCUSLJVVRCA-UHFFFAOYSA-M
Synonym 4-thioanisolemagnesium bromide, 4-methylthio phenylmagnesium bromide, 4-thioanisolemagnesium bromide solution, ssvqcusljvvrca-uhfffaoysa-m, 4-methylsulfanylphenylmagnesium bromide, 4-methylthio phenyl magnesium bromide, 4-methylsulfanyl-phenylmagnesium bromide, magnesium,bromo 4-methylthio phenyl, 4-thioanisolemagnesium bromide 0.5 m in tetrahydrofuran, 4-thioanisolemagnesium bromide solution, 0.5 m in thf, grignard reagent
PubChem CID 10857186
IUPAC Name magnesium;methylsulfanylbenzene;bromide
SMILES CSC1=CC=C([Mg]Br)C=C1

Specifications

CAS Min % 9.43, 85.86
CAS Max % 14.14, 90.57
Color Yellow to Brown
Density 0.9650g/mL
Flash Point <−17°C
Packaging AcroSeal™ Glass Bottle
Linear Formula CH3SC6H4MgBr
Quantity 100 mL
Specific Gravity 0.965
Solubility Information Solubility in water: reacts.
Formula Weight 227.4
Physical Form Solution
Chemical Name or Material 4-Thioanisolemagnesium bromide, 0.5M solution in THF
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
Recommended Storage Product may develop some turbidity or precipitate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.