accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18622-23-6

NH2HNO

CAS RN 18622-23-6

IUPAC Name: [1,1'-biphenyl]-4-carbohydrazide
Synonyms: [1,1'-biphenyl]-4-carbohydrazide
Molecular Weight (g/mol): 212.25
Molecular Formula: C13H12N2O
InChi Key: QEUAQXSDDNDOTG-UHFFFAOYSA-N
SMILES: NNC(=O)C1=CC=C(C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results