Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-biphenylcarboxylic acid hydrazide

Catalog No. CDS014638 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS014638 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS014638 Supplier Sigma Aldrich Supplier No. CDS014638100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Synthesis of (trimethylsilyl)methyl isocyanide and its reaction with protic nucleophiles has been reported
  • An improved preparation of (trimethylsilyl)methyl isocyanide is reported
  • Reaction of (trimethylsilyl)methyl isocyanide with (++2-formaldehyde)zirconocene complex is reported to yield insertion products.(trimethylsilyl)methyl isocyanide may be used in the preparation of isocyanomethylenetriphenylphosphorane

Specifications

CAS 18622-23-6
Molecular Formula C13H12N2O
Linear Formula C13H12N2O
MDL Number MFCD00017078
Quantity 500 mg
Molecular Weight (g/mol) 212.25
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.