accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18656-96-7

IUPAC Name: cyclohexanaminium 6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
Synonyms: cyclohexylammonium ion 6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate
Molecular Weight (g/mol): 521.79
Molecular Formula: C20H26BrClN2O7
InChi Key: JXCKZXHCJOVIAV-UHFFFAOYNA-N
SMILES: [NH3+]C1CCCCC1.OC1C(O)C(OC2=CNC3=CC=C(Br)C(Cl)=C23)OC(C1O)C([O-])=O
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results