accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 186650-98-6

CH3H3CCH3OONNN

CAS RN 186650-98-6

IUPAC Name: tert-butyl 4-(4-cyanophenyl)piperazine-1-carboxylate
Synonyms: tert-butyl 4-(4-cyanophenyl)piperazine-1-carboxylate
Molecular Weight (g/mol): 287.36
Molecular Formula: C16H21N3O2
InChi Key: WIBWVYVIBJOVIW-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCN(CC1)C1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results