accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3OCH

CAS RN 18669-04-0

IUPAC Name: 1-ethoxy-1-(prop-2-yn-1-yloxy)ethane
Synonyms: 1-ethoxy-1-(prop-2-yn-1-yloxy)ethane
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: QKBBTQJLUGADEG-UHFFFAOYNA-N
SMILES: CCOC(C)OCC#C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results