accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFOFFF

CAS RN 1868-00-4

IUPAC Name: bis[3-(trifluoromethyl)phenyl]methanone
Synonyms: bis[3-(trifluoromethyl)phenyl]methanone
Molecular Weight (g/mol): 318.22
Molecular Formula: C15H8F6O
InChi Key: POOXOHISLGOAEF-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=CC=C1)C(=O)C1=CC(=CC=C1)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results