accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1868-53-7

IUPAC Name: dibromo(fluoro)methane
Synonyms: dibromofluoromethane
Molecular Weight (g/mol): 191.83
Molecular Formula: CHBr2F
InChi Key: LTUTVFXOEGMHMP-UHFFFAOYSA-N
SMILES: FC(Br)Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results