accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SOOOH3COOH3CIClClOCH3

CAS RN 1868173-25-4

IUPAC Name: (3,5-dichlorophenyl)(2,4,6-trimethoxyphenyl)iodanium 4-methylbenzene-1-sulfonate
Synonyms: (3,5-dichlorophenyl)(2,4,6-trimethoxyphenyl)iodanium tosylate
Molecular Weight (g/mol): 611.27
Molecular Formula: C22H21Cl2IO6S
InChi Key: AYHVNGRMSLNAJQ-UHFFFAOYSA-M
SMILES: CC1=CC=C(C=C1)S([O-])(=O)=O.COC1=CC(OC)=C([I+]C2=CC(Cl)=CC(Cl)=C2)C(OC)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results