CAS RN 18694-40-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Mepirizole, MilliporeSigma™ Supelco™
Mepirizole belongs to the class of non-steroidal anti-inflammatory drugs (NSAIDs), widely used in the treatment of pain and inflammation.