Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mepirizole, MilliporeSigma™ Supelco™

Catalog No. 111013398 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3398 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3398 Supplier MilliporeSigma Supelco Supplier No. M901710G
Add to Cart
Add to Cart

Mepirizole belongs to the class of non-steroidal anti-inflammatory drugs (NSAIDs), widely used in the treatment of pain and inflammation.

TRUSTED_SUSTAINABILITY

Specifications

CAS 18694-40-1
Physical Form Neat
Linear Formula C11H14N4O2
MDL Number MFCD00057351
Type Analytical Standard
Quantity 10 g
Formula Weight 234.25
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.