accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOSNHOOClClClOOCH3OOCH3

CAS RN 187022-49-7

IUPAC Name: [3,4-bis(acetyloxy)-6-(phenylsulfanyl)-5-{[(2,2,2-trichloroethoxy)carbonyl]amino}oxan-2-yl]methyl acetate
Synonyms: [3,4-bis(acetyloxy)-6-(phenylsulfanyl)-5-{[(2,2,2-trichloroethoxy)carbonyl]amino}oxan-2-yl]methyl acetate
Molecular Weight (g/mol): 572.83
Molecular Formula: C21H24Cl3NO9S
InChi Key: AZDNCEYOFMNMSH-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(SC2=CC=CC=C2)C(NC(=O)OCC(Cl)(Cl)Cl)C(OC(C)=O)C1OC(C)=O
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)