accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3COOOH3CNHOO

CAS RN 1872-59-9

IUPAC Name: methyl 2-{[(benzyloxy)carbonyl]amino}-3-(tert-butoxy)propanoate
Synonyms: methyl 2-{[(benzyloxy)carbonyl]amino}-3-(tert-butoxy)propanoate
Molecular Weight (g/mol): 309.36
Molecular Formula: C16H23NO5
InChi Key: UMAXPCYVVRYQNO-UHFFFAOYNA-N
SMILES: COC(=O)C(COC(C)(C)C)NC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results