Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

O-tert-Butyl-N-carbobenzoxy-L-serine Methyl Ester 98.0+%, TCI America™
SDP

Catalog No. B17325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17325G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B17325G Supplier TCI America Supplier No. B17325G

Chemical Identifiers

CAS 1872-59-9
Molecular Formula C16H23NO5
Molecular Weight (g/mol) 309.362
MDL Number MFCD00191072
InChI Key UMAXPCYVVRYQNO-ZDUSSCGKSA-N
Synonym O-tert-Butyl-N-Cbz-L-serine Methyl Ester, Z-Ser(tBu)-OMe
PubChem CID 14629315
IUPAC Name methyl (2S)-3-[(2-methylpropan-2-yl)oxy]-2-(phenylmethoxycarbonylamino)propanoate
SMILES CC(C)(C)OCC(C(=O)OC)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 43°C
Color White-Yellow
Quantity 5 g
Formula Weight 309.36
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material O-tert-Butyl-N-carbobenzoxy-L-serine Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.