accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SiClClH2C

CAS RN 1873-92-3

IUPAC Name: dichloro(methyl)(prop-2-en-1-yl)silane
Synonyms: dichloro(methyl)prop-2-en-1-ylsilane
Molecular Weight (g/mol): 155.09
Molecular Formula: C4H8Cl2Si
InChi Key: YCEQUKAYVABWTE-UHFFFAOYSA-N
SMILES: C[Si](Cl)(Cl)CC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results