accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOHNNHNOOCH3CH3OCH3(Z)(Z)

CAS RN 187393-00-6

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: —
SMILES: —
Grade 
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)