Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bemotrizinol, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013401 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3401 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3401 Supplier MilliporeSigma Supelco Supplier No. 5182525MG
Add to Cart
Add to Cart
Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 187393-00-6
Physical Form Neat
Linear Formula C38H49N3O5
MDL Number MFCD08435974
Melting Point 85°C to 87°C
Synonym 2,4-Bis[4-(2-ethylhexyloxy)-2-hydroxyphenyl]-6-(4-methoxyphenyl)-1,3,5-triazine
Type Analytical Standard
Quantity 25 mg
Formula Weight 627.81
Percent Purity ≥98% (HPLC)
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) hydrogen content, 7.87% (theory)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.