accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOOO(E)

CAS RN 1874-22-2

IUPAC Name: (2E)-3-(5-nitrofuran-2-yl)prop-2-enal
Synonyms: (2E)-3-(5-nitrofuran-2-yl)prop-2-enal
Molecular Weight (g/mol): 167.12
Molecular Formula: C7H5NO4
InChi Key: DXWCZMGIIFEEPU-OWOJBTEDSA-N
SMILES: [O-][N+](=O)C1=CC=C(O1)\C=C\C=O
Grade 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results