accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18744-58-6

IUPAC Name: barium(2+) 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
Synonyms: barium(2+) ion(4-) edta
Molecular Weight (g/mol): 425.54
Molecular Formula: C10H12BaN2O8
InChi Key: MASKRDCJSCCWNE-UHFFFAOYSA-J
SMILES: [Ba++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results