Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Barium Disodium Ethylenediaminetetraacetate Hydrate 98.0+%, TCI America™
SDP

Catalog No. E008525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E008525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E008525G Supplier TCI America Supplier No. E008525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chelation reagent with a purity of at least 98.0% by chelometric titration.

Chemical Identifiers

CAS 18744-58-6
Molecular Formula C10H12BaN2O8
Molecular Weight (g/mol) 425.54
MDL Number MFCD00054444
InChI Key MASKRDCJSCCWNE-UHFFFAOYSA-J
Synonym Ethylenediaminetetraacetic Acid Disodium Barium Salt, Disodium Barium Ethylenediaminetetraacetate, Ethylenediaminetetraacetic Acid Barium Disodium Salt
PubChem CID 102601892
IUPAC Name barium(2+) 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
SMILES [Ba++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

Color White
pH 6.5 at 8.0 (5% H2O soln.)
Quantity 25 g
UN Number 1564
Formula Weight 471.52
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Barium Disodium Ethylenediaminetetraacetate Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.