accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18746-63-9

IUPAC Name: λ⁴-ruthenium(4+) diammonium hexachloride
Synonyms: λ⁴-ruthenium(4+) diammonium hexachloride
Molecular Weight (g/mol): 349.85
Molecular Formula: Cl6H8N2Ru
InChi Key: DHTSBQSPUBOKKI-UHFFFAOYSA-J
SMILES: [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru+4]
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)