Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium hexachlororuthenate(IV), Ru 28.4% min

Catalog No. AA1187406
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1187406 5 g
AA1187403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1187406 Supplier Thermo Scientific Chemicals Supplier No. 01187406
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 18746-63-9 | Cl6H8N2Ru | 349.85 g/mol

Ammonium hexachlororuthenate(IV) is used in laboratory chemicals and manufacture of other substances. Ammonium hexachlororuthenate (ACR) is a salt precipitated during a substitution reaction between ammonium and ruthenate dissolved in a strong hydrochloric acid medium. This precipitation reaction is used within the platinum industry as a means of recovering and purifying ruthenium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18746-63-9
Molecular Formula Cl6H8N2Ru
Molecular Weight (g/mol) 349.85
MDL Number MFCD00015962
InChI Key DHTSBQSPUBOKKI-UHFFFAOYSA-J
Synonym ammonium hexachlororuthenate iv, diammonium hexachlororuthenate, ammonium chlororuthenate, ammonium hexachlororuthenate, epitope id:151520, ruthenium iv-ammonium chloride, ammonium hexachloridoruthenate iv, ammonium hexachloridoruthenate 2-, diammonium hexachlororuthenate 2-
PubChem CID 46173598
ChEBI CHEBI:60134
SMILES [NH4+].[NH4+].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru+4]

Specifications

Color Red to Brown
Physical Form Crystalline Powder
Quantity 5 g
Solubility Information Soluble in water.
IUPAC Name λ⁴-ruthenium(4+) diammonium hexachloride
Formula Weight 349.87
Concentration or Composition (by Analyte or Components) Ru 28.4% min.
Odor Odorless
Sensitivity Hygroscopic
Chemical Name or Material Ammonium hexachlororuthenate(IV)
Hazard Category H301+H311+H331
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 242-552-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.