accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 187731-26-6

IUPAC Name: hexyldimethyloctylazanium bromide
Synonyms: hexyldimethyloctylazanium bromide
Molecular Weight (g/mol): 322.38
Molecular Formula: C16H36BrN
InChi Key: KIEOLZRKTOBVMX-UHFFFAOYSA-M
SMILES: [Br-].CCCCCCCC[N+](C)(C)CCCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results