Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexyldimethyloctylammonium Bromide 97.0+%, TCI America™
SDP

Catalog No. H09895G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09895G 5 g
H098925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09895G Supplier TCI America Supplier No. H09895G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 187731-26-6
Molecular Formula C16H36BrN
Molecular Weight (g/mol) 322.38
MDL Number MFCD03093632
InChI Key KIEOLZRKTOBVMX-UHFFFAOYSA-M
PubChem CID 44630344
IUPAC Name hexyldimethyloctylazanium bromide
SMILES [Br-].CCCCCCCC[N+](C)(C)CCCCCC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 322.38
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material Hexyldimethyloctylammonium Bromide
EINECSNumber (9)-1971
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.