accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1878-69-9

IUPAC Name: 2-(3-iodophenyl)acetic acid
Synonyms: M-iodophenylacetic acid
Molecular Weight (g/mol): 262.05
Molecular Formula: C8H7IO2
InChi Key: MRSWWBAFGGGWRH-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CC(I)=CC=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)