Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Iodophenylacetic acid, 98%, Thermo Scientific™

Catalog No. AC443630010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC443630010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC443630010 Supplier Thermo Scientific Chemicals Supplier No. 443630010

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1878-69-9
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.05
MDL Number MFCD00046548
InChI Key MRSWWBAFGGGWRH-UHFFFAOYSA-N
Synonym 3-iodophenylacetic acid, 2-3-iodophenyl acetic acid, 3-iodophenyl acetic acid, m-iodophenylacetic acid, m-iodobenzeneacetic acid, benzeneacetic acid, 3-iodo, pubchem4195, 3-iodophenylaceticacid, acmc-1cfam, 3-iodobenzeneacetic acid
PubChem CID 3870220
IUPAC Name 2-(3-iodophenyl)acetic acid
SMILES OC(=O)CC1=CC(I)=CC=C1

Specifications

Melting Point 130.0°C to 132.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 262.05
Percent Purity 98%
Chemical Name or Material 3-Iodophenylacetic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.