accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1878-91-7

IUPAC Name: 2-(4-bromophenoxy)acetic acid
Synonyms: P-bromophenoxyacetic acid
Molecular Weight (g/mol): 231.05
Molecular Formula: C8H7BrO3
InChi Key: SZEBGAQWWSUOHT-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results