Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromophenoxyacetic Acid 98.0+%, TCI America™
SDP

Catalog No. B27465G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27465G 5 g
B274625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27465G Supplier TCI America Supplier No. B27465G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1878-91-7
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.045
InChI Key SZEBGAQWWSUOHT-UHFFFAOYSA-N
Synonym 4-bromophenoxyacetic acid, 2-4-bromophenoxy acetic acid, 4-bromophenoxy acetic acid, p-bromophenoxyacetic acid, acetic acid, 4-bromophenoxy, p-bromophenoxyacetate, p-bromophenoxyacetic acvid, acetic acid, 2-4-bromophenoxy, acetic acid, p-bromophenoxy
PubChem CID 74657
IUPAC Name 2-(4-bromophenoxy)acetic acid
SMILES C1=CC(=CC=C1OCC(=O)O)Br

Specifications

Melting Point 159°C
Color White-Yellow
Quantity 5 g
Formula Weight 231.05
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromophenoxyacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.