CAS RN 187863-42-9
IUPAC Name:
1-butylpyridin-1-ium; trifluoro[(trifluoromethanesulfonylazanidyl)sulfonyl]methane
Synonyms:
1-butylpyridin-1-ium; bistriflylimide anion
Molecular Weight (g/mol):
416.35
Molecular Formula:
C11H14F6N2O4S2
InChi Key:
XHIHMDHAPXMAQK-UHFFFAOYSA-N
SMILES:
CCCC[N+]1=CC=CC=C1.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F
1
–
1
of
1
results
1
1-Butylpyridinium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
CAS: 187863-42-9 Molecular Formula: C11H14F6N2O4S2 Molecular Weight (g/mol): 416.353 MDL Number: MFCD16038143 InChI Key: XHIHMDHAPXMAQK-UHFFFAOYSA-N PubChem CID: 12184310 IUPAC Name: bis(trifluoromethylsulfonyl)azanide;1-butylpyridin-1-ium SMILES: CCCC[N+]1=CC=CC=C1.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F