Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Butylpyridinium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
SDP

Catalog No. B55715G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55715G 5 g
B557125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55715G Supplier TCI America Supplier No. B55715G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 187863-42-9
Molecular Formula C11H14F6N2O4S2
Molecular Weight (g/mol) 416.353
MDL Number MFCD16038143
InChI Key XHIHMDHAPXMAQK-UHFFFAOYSA-N
PubChem CID 12184310
IUPAC Name bis(trifluoromethylsulfonyl)azanide;1-butylpyridin-1-ium
SMILES CCCC[N+]1=CC=CC=C1.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 416.35
Percent Purity ≥98.0% (HPLC,T)
Grade Research
Physical Form Liquid
Chemical Name or Material 1-Butylpyridinium Bis(trifluoromethanesulfonyl)imide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.