accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 18800-53-8

IUPAC Name: (3,4-dimethylphenyl)sulfanide
Synonyms: (3,4-dimethylphenyl)sulfanide
Molecular Weight (g/mol): 137.22
Molecular Formula: C8H9S
InChi Key: IDKCKPBAFOIONK-UHFFFAOYSA-M
SMILES: CC1=CC=C([S-])C=C1C
Boiling Point 
  • (5)
  • (1)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (4)
  • (5)
  • (1)
  • (4)
  • (2)
  • (1)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)