Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,4-Dimethylbenzenethiol 97.0+%, TCI America™
SDP

Catalog No. D334425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D334425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D334425G Supplier TCI America Supplier No. D334425G

missing translation for 'validMainframeMsg'

Specifications

CAS 18800-53-8
Color Yellow
Boiling Point 218°C
Molecular Formula C8H10S
Quantity 25 g
Synonym 3,4-dimethylthiophenol, benzenethiol, 3,4-dimethyl, 3,4-dimethyl thiophenol, o-xylene-4-thiol, 4-mercapto-o-xylene, 3,4-dimethylbenzene-1-thiol, pubchem6812, 3,4-dimethyl-thiophenol, acmc-1bu1r, 3,4-dimethyl-benzenethiol
InChI Key IDKCKPBAFOIONK-UHFFFAOYSA-N
SMILES CC1=C(C=C(C=C1)S)C
IUPAC Name 3,4-dimethylbenzenethiol
Molecular Weight (g/mol) 138.228
PubChem CID 87799
Formula Weight 138.23
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3,4-Dimethylbenzenethiol
Show More Show Less
EINECSNumber (3)-1094
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.