accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3C

CAS RN 18801-00-8

IUPAC Name: 2-tert-butylanthracene
Synonyms: 2-(tert-butyl)anthracene
Molecular Weight (g/mol): 234.34
Molecular Formula: C18H18
InChi Key: WBPXZSIKOVBSAS-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C2C=C3C=CC=CC3=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results