Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-tert-Butylanthracene 98.0+%, TCI America™
SDP

Catalog No. B35415G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35415G 5 g
B35411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35415G Supplier TCI America Supplier No. B35415G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18801-00-8
Molecular Formula C18H18
Molecular Weight (g/mol) 234.342
MDL Number MFCD00003581
InChI Key WBPXZSIKOVBSAS-UHFFFAOYSA-N
PubChem CID 87800
IUPAC Name 2-tert-butylanthracene
SMILES CC(C)(C)C1=CC2=CC3=CC=CC=C3C=C2C=C1

Specifications

Melting Point 148°C
Color White-Yellow
Quantity 5 g
Synonym 2-tert-butyl anthracene, anthracene, 2-1,1-dimethylethyl, anthracene,2-1,1-dimethylethyl, 2-tertbutylanthracene, 2-t-butyl anthracene, acmc-209eqy
Formula Weight 234.34
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-tert-Butylanthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.