accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COFMgBr

CAS RN 188132-02-7

IUPAC Name: bromo(5-fluoro-2-methoxyphenyl)magnesium
Synonyms: bromo(5-fluoro-2-methoxyphenyl)magnesium
Molecular Weight (g/mol): 229.33
Molecular Formula: C7H6BrFMgO
InChi Key: ZYVBZHZWABCRCW-UHFFFAOYSA-M
SMILES: COC1=CC=C(F)C=C1[Mg]Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results