Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-2-methoxyphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

Catalog No. AC431880500
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431880500 50 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC431880500 Supplier Thermo Scientific Chemicals Supplier No. 431880500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 188132-02-7 | C7H6BrFMgO | 229.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 188132-02-7 , 109-99-9
Molecular Formula C7H6BrFMgO
Molecular Weight (g/mol) 229.33
MDL Number MFCD01311481
InChI Key ZYVBZHZWABCRCW-UHFFFAOYSA-M
Synonym magnesium 1-fluoro-4-methoxybenzene-5-ide bromide, 5-fluoro-2-methoxyphenylmagnesium bromide solution, magnesium bromide 5-fluoro-2-methoxybenzen-1-ide 1/1/1, magnesium 2+ ion 5-fluoro-2-methoxybenzen-1-ide bromide, 5-fluoro-2-methoxyphenylmagnesium bromide 0.5 m solution in thf, spcseal, grignard reagent
PubChem CID 2778879
IUPAC Name magnesium;1-fluoro-4-methoxybenzene-5-ide;bromide
SMILES COC1=CC=C(F)C=C1[Mg]Br

Specifications

CAS Min % 9.68, 85.49
CAS Max % 14.51, 90.32
Color Yellow to Brown
Density 0.9480g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Linear Formula F(OCH3)C6H3MgBr
Quantity 50 mL
Specific Gravity 0.948
Solubility Information Solubility in water: reacts
Formula Weight 229.33
Physical Form Solution
Chemical Name or Material 5-Fluoro-2-methoxyphenylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.