accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3NNOCH3H3CCH3CH3CH3H3COH3CH3CCH3Co(E/Z)(E/Z)

CAS RN 188264-84-8

IUPAC Name: λ²-cobalt(2+) 2,4-di-tert-butyl-6-{[(2-{[(3,5-di-tert-butyl-2-oxidophenyl)methylidene]amino}cyclohexyl)imino]methyl}benzen-1-olate
Synonyms: λ²-cobalt(2+) 2,4-di-tert-butyl-6-{[(2-{[(3,5-di-tert-butyl-2-oxidophenyl)methylidene]amino}cyclohexyl)imino]methyl}benzenolate
Molecular Weight (g/mol): 603.76
Molecular Formula: C36H52CoN2O2
InChi Key: ZFWPDJKMASHRPT-UHFFFAOYNA-L
SMILES: [Co++].CC(C)(C)C1=CC(=C([O-])C(C=NC2CCCCC2N=CC2=CC(=CC(=C2[O-])C(C)(C)C)C(C)(C)C)=C1)C(C)(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results