accessibility menu, dialog, popup

UserName

Viewing profile as user:
OMoO

CAS RN 18868-43-4

IUPAC Name: dioxomolybdenum
Synonyms: molybdenum(IV) oxide
Molecular Weight (g/mol): 127.95
Molecular Formula: MoO2
InChi Key: QXYJCZRRLLQGCR-UHFFFAOYSA-N
SMILES: O=[Mo]=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)