Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Molybdenum(IV) oxide

Catalog No. 23476150G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-476-150G 50 g
23-476-110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-476-150G Supplier Sigma Aldrich Supplier No. 23476150G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Molybdenum(IV) oxide (MoO2) is a transition metal oxide with good electrical conductivity due to the presence of delocalized electrons in its valence band. It is traditionally prepared by reducing MoO3 in dry hydrogen in the temperature range of 450°C to 550°C.MoO2 can be used in a variety of energy based applications such as an anode material in rechargeable lithium batteries and fuel flexible solid oxide fuel cells. It can also be used as a catalyst in hydrogen reforming processes.

Specifications

CAS 18868-43-4
Quantity 50 g
Molecular Formula MoO2
Linear Formula MoO2
MDL Number MFCD00014223
Molecular Weight (g/mol) 127.94
Percent Purity 99%
Density 6.47 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.