CAS RN 18869-30-2
IUPAC Name:
1-bromo-4-[2-(4-bromophenyl)ethenyl]benzene
Synonyms:
1-bromo-4-[2-(4-bromophenyl)ethenyl]benzene
Molecular Weight (g/mol):
338.04
Molecular Formula:
C14H10Br2
InChi Key:
JEHMPNUQSJNJDL-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(C=CC2=CC=C(Br)C=C2)C=C1
1
–
1
of
1
results
1
4,4'-Dibromo-trans-stilbene 98.0+%, TCI America™
CAS: 18869-30-2 Molecular Formula: C14H10Br2 Molecular Weight (g/mol): 338.04 MDL Number: MFCD00448009 InChI Key: JEHMPNUQSJNJDL-UHFFFAOYSA-N PubChem CID: 7020566 IUPAC Name: 1-bromo-4-[2-(4-bromophenyl)ethenyl]benzene SMILES: BrC1=CC=C(C=CC2=CC=C(Br)C=C2)C=C1