Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dibromo-trans-stilbene 98.0+%, TCI America™
SDP

Catalog No. D30435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D30435G 5 g
D3043250MG 250 mg
D30431G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D30435G Supplier TCI America Supplier No. D30435G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18869-30-2
Molecular Formula C14H10Br2
Molecular Weight (g/mol) 338.04
MDL Number MFCD00448009
InChI Key JEHMPNUQSJNJDL-UHFFFAOYSA-N
PubChem CID 7020566
IUPAC Name 1-bromo-4-[2-(4-bromophenyl)ethenyl]benzene
SMILES BrC1=CC=C(C=CC2=CC=C(Br)C=C2)C=C1

Specifications

Melting Point 212°C
Color White-Yellow
Quantity 5 g
Formula Weight 338.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dibromo-trans-stilbene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.