accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1887126-77-3

IUPAC Name: 3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Synonyms: 3-fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Molecular Weight (g/mol): 238.07
Molecular Formula: C12H16BFO3
InChi Key: KBGOBMOAZQOCTA-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(O)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results